missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromohexadecane-1,1,2,2-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Bromohexadecane-1,1,2,2-D4 |
| CAS | 1219805-65-8 |
| Molecular Formula | C16 D4 H29 Br |
| Quantity | 0.1 g |
| Synonym | 1-Bromohexadecane-1,1,2,2-d4, 1-Hexadecyl-1,1,2,2-d4 bromide, Cetyl-1,1,2,2-d4 bromide |
| InChI Formula | InChI=1 S/C16H33Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h2-16H2,1H3/i15D2,16D2 |
| SMILES | [2 H]C([2 H])(Br)C([2 H])([2 H])CCCCCCCCCCCCCC |
| IUPAC Name | 1-bromo-1,1,2,2-tetradeuteriohexadecane |
| Molecular Weight (g/mol) | 309.3619 |
| Formula Weight | 308.2017 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction