missing translation for 'onlineSavingsMsg'
Learn More

1-Bromohexadecane, 97%

Product Code. 10194500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10194500 1 L Glass bottle 1 L
10104510 250 mL Glass bottle 250mL
11984491 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10194500 Supplier Thermo Scientific Acros Supplier No. 215430010
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 112-82-3 | C16H33Br | 305.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-82-3
Molecular Formula C16H33Br
Molecular Weight (g/mol) 305.34
MDL Number MFCD00000230
InChI Key HNTGIJLWHDPAFN-UHFFFAOYSA-N
Synonym hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam
PubChem CID 8213
IUPAC Name 1-bromohexadecane
SMILES CCCCCCCCCCCCCCCCBr

Specifications

Melting Point 17.3°C
Color Amber to Black-Brown
Density 0.9990g/mL
Boiling Point 336.0°C
Flash Point 177°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)15Br
Refractive Index 1.4609 to 1.4629
Quantity 1 L
Beilstein 01,172
Specific Gravity 0.999
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 305.33
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material 1-Bromohexadecane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.