missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromohexane-2,2,3,3,4,4,5,5,6,6,6-d11, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromohexane-2,2,3,3,4,4,5,5,6,6,6-D11 |
| CAS | 2159-17-3 |
| Molecular Formula | C6 D11 H2 Br |
| Quantity | 0.5 g |
| Synonym | Hexane-1,1,1,2,2,3,3,4,4,5,5-d11, 6-bromo- (7 CI, 9 CI), 6-Bromohexane-1,1,1,2,2,3,3,4,4,5,5-d11, 1-Bromohexane-2,2,3,3,4,4,5,5,6,6,6-d11, 1-Bromohexane-d11, 1-Bromohexane-2,2,3,3,4,4,5,5,6,6,6-D11 |
| InChI Formula | InChI=1 S/C6H13Br/c1-2-3-4-5-6-7/h2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CBr |
| IUPAC Name | 6-bromo-1,1,1,2,2,3,3,4,4,5,5-undecadeuteriohexane |
| Molecular Weight (g/mol) | 176.1392 |
| Formula Weight | 175.0891 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction