missing translation for 'onlineSavingsMsg'
Learn More

1-Bromohexane, 99+%

Product Code. 11904491 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
10 L
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 11904491

Brand: Thermo Scientific Acros 107040100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-25-1 | C6H13Br | 165.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-25-1
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.07
MDL Number MFCD00000271
InChI Key MNDIARAMWBIKFW-UHFFFAOYSA-N
Synonym hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga
PubChem CID 8101
IUPAC Name 1-bromohexane
SMILES CCCCCCBr

Specifications

Acidity 0.1% max. (as HBr)
CAS Min % 99.0
CAS Max % 100.0
Melting Point -85°C
Density 1.1700g/mL
Boiling Point 154°C to 158°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 0.5% max. Hexanol (GC)
Packaging Plastic drum
Linear Formula CH3(CH2)5Br
Refractive Index 1.4465 to 1.4485
Quantity 10 L
Beilstein 01,144
Specific Gravity 1.17
Solubility Information Solubility in water: insoluble
Formula Weight 165.07
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1-Bromohexane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Bromohexane
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.