missing translation for 'onlineSavingsMsg'
Learn More

1-Bromomethyl-2,2-difluorocyclopropane, 97%

Product Code. 15467247
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467247 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15467247 Supplier Thermo Scientific Alfa Aesar Supplier No. H34305.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77613-65-1 | C4H5BrF2 | 170.985 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 77613-65-1
Boiling Point 105°C to 107°C
Molecular Formula C4H5BrF2
MDL Number MFCD06797749
Quantity 250 mg
UN Number UN1992
Synonym 1-bromomethyl-2,2-difluorocyclopropane, 2-bromomethyl-1,1-difluorocyclopropane, 2,2-difluorocyclopropylmethyl bromide, 2,2-difluorocycloprop-1-yl methyl bromide, bromomethyl-2,2-difluorocyclopropane, 1-bromomethyl-2,2-difluoro-cyclopropane, 2-brornomethyl-1,1-difluorocyclopropane, cyclopropane,2-bromomethyl-1,1-difluoro, cyclopropane, 2-bromomethyl-1,1-difluoro, 2-bromomethyl-1,1-bis fluoranyl cyclopropane
InChI Key TUDJNSKRXIUOAJ-UHFFFAOYSA-N
SMILES C1C(C1(F)F)CBr
IUPAC Name 2-(bromomethyl)-1,1-difluorocyclopropane
Molecular Weight (g/mol) 170.985
PubChem CID 2761402
Formula Weight 170.98
Percent Purity 97%
Chemical Name or Material 1-Bromomethyl-2,2-difluorocyclopropane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.