missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)-2,3,4,5,6-pentafluorobenzene, 97%, BLD Pharm™

Product Code. 30801826
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30801826 25 g 25g
30793849 5 g 5g
30787097 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30801826 Supplier BLD Pharm Supplier No. BD14701525g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1765-40-8
Molecular Formula C7H2BrF5
MDL Number MFCD00000299
Quantity 25 g
SMILES FC1=C(CBr)C(F)=C(F)C(F)=C1F
Molecular Weight (g/mol) 260.99
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)-2,3,4,5,6-pentafluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.