missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)adamantane, 97%, Thermo Scientific™

Product Code. 10509852
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10509852 1 g Amber Glass Bottle 1g
10396494 5 g Amber Glass Bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10509852

Brand: Thermo Scientific Maybridge BTB13836DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 14651-42-4
Molecular Formula C11H17Br
Molecular Weight (g/mol) 229.161
InChI Key RLRBYCAVZBQANU-UHFFFAOYSA-N
Synonym 1-bromomethyl adamantane, tricyclo 3.3.1.13,7 decane, 1-bromomethyl, adamantanylbromomethane, 1-bromomethyl tricyclo 3.3.1.1∼3,7∼ decane, 3r,5s,7s-1-bromomethyl adamantane, pubchem21524, 1-adamantylmethylbromide, 1-bromoethyl adamantane, 1-adamantyl methyl bromide, 1s,3r,5s,7s-1-bromomethyl adamantane
PubChem CID 1416304
IUPAC Name 1-(bromomethyl)adamantane
SMILES C1C2CC3CC1CC(C2)(C3)CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 229.16
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)adamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.