missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)cyclopropane, 95%, Thermo Scientific™

Product Code. 10258242
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10258242 25 g Amber Glass Bottle 25g
10205904 1 g Amber Glass Bottle 1g
10080062 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10258242 Supplier Thermo Scientific Maybridge Supplier No. RJF00467EB
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7051-34-5
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.00
MDL Number MFCD00001306
InChI Key AEILLAXRDHDKDY-UHFFFAOYSA-N
Synonym bromomethyl cyclopropane, cyclopropylmethyl bromide, cyclopropane, bromomethyl, cyclopropyl bromo methane, 1-bromomethyl cyclopropane, bromocyclopropylmethane, cyclopropylmethylbromide, bromomethyl-cyclopropane, cyclopropyl methylbromide
PubChem CID 81503
SMILES BrCC1CC1

Specifications

CAS Min % 95.0
Density 1.3800g/mL
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula C3H5CH2Br
Quantity 25 g
IUPAC Name bromomethylcyclopropane
Formula Weight 135
Percent Purity 95%
Chemical Name or Material 1-(Bromomethyl)cyclopropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.