missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromononane-d19, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Bromononane-D19 |
| CAS | 1219805-90-9 |
| Molecular Formula | C9 D19 Br |
| Quantity | 0.25 g |
| Synonym | Nonane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-d19, 9-bromo- (ACI), 9-Bromononane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-d19 (ACI), 1-Bromononane-d19, 1-Bromononane-D19, 9-Bromononane-d19, 1-Nonyl-d19 bromide |
| InChI Formula | InChI=1 S/C9H19Br/c1-2-3-4-5-6-7-8-9-10/h2-9H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecadeuteriononane |
| Molecular Weight (g/mol) | 226.2682 |
| Formula Weight | 225.1863 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction