missing translation for 'onlineSavingsMsg'
Learn More

1-Bromooctadecane, 96%

Product Code. 15179213 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15179213 2.5 kg Glass bottle 2.5kg
10467750 5 g Glass bottle 5g
10591441 100 g Glass bottle 100g
10407760 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15179213 Supplier Thermo Scientific Acros Supplier No. 190020025
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
This item is not returnable. View return policy

CAS: 112-89-0 | C18H37Br | 333.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-89-0
Molecular Formula C18H37Br
Molecular Weight (g/mol) 333.40
MDL Number MFCD00000231
InChI Key WSULSMOGMLRGKU-UHFFFAOYSA-N
Synonym octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane
PubChem CID 8218
IUPAC Name 1-bromooctadecane
SMILES CCCCCCCCCCCCCCCCCCBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point >20.0°C
Color Yellow
Density 0.9700g/mL
Boiling Point 214.0°C to 216.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)17Br
Refractive Index 1.4620 to 1.4640
Quantity 2.5 kg
Beilstein 01,I,69
Specific Gravity 0.97
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 333.38
Percent Purity 96%
Physical Form Low Melting Solid
Chemical Name or Material 1-Bromooctadecane, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.