missing translation for 'onlineSavingsMsg'
Learn More

1-Bromooctane, 99%

Product Code. 10071910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10071910 500 mL Glass bottle 500mL
15740819 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10071910 Supplier Thermo Scientific Acros Supplier No. 166815000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111-83-1 | C8H17Br | 193.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-83-1
Molecular Formula C8H17Br
Molecular Weight (g/mol) 193.13
MDL Number MFCD00000276
InChI Key VMKOFRJSULQZRM-UHFFFAOYSA-N
Synonym n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo
PubChem CID 8140
IUPAC Name 1-bromooctane
SMILES CCCCCCCCBr

Specifications

Melting Point -55.0°C
Density 1.1100g/mL
Boiling Point 201.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)7Br
Refractive Index 1.4508 to 1.4528
Quantity 500 mL
Beilstein 01,16
Merck Index 15,6853
Specific Gravity 1.11
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Formula Weight 193.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Bromooctane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.