missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoperfluoroheptane, 97%

Product Code. 11469432
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469432 25 g 25g
11449432 1 g 1g
11459432 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469432 Supplier Thermo Scientific Alfa Aesar Supplier No. A10156.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 375-88-2 | C7BrF15 | 448.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-88-2
Molecular Formula C7BrF15
Molecular Weight (g/mol) 448.96
MDL Number MFCD00013570
InChI Key VPQQZKWYZYVTMU-UHFFFAOYSA-N
Synonym 1-bromoperfluoroheptane, perfluoroheptyl bromide, 1-bromopentadecafluoroheptane, perfluoro-n-heptyl bromide, pentadecafluoroheptyl bromide, heptane, 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoro, acmc-1ad35, 1-bromopentadecafluoroheptane; perfluoroheptyl bromide, 1-bromanyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecakis fluoranyl heptane
PubChem CID 67819
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br

Especificaciones

Density 1.894
Boiling Point 117°C to 119°C
Flash Point None
Linear Formula CF3(CF2)6Br
Refractive Index 1.303
Quantity 25 g
Beilstein 1716336
Formula Weight 448.96
Percent Purity 97%
Chemical Name or Material 1-Bromoperfluoroheptane

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.