missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromopropane-1-d1, CDN
missing translation for 'orderingAttributeHoverText'
Quantity:
0.5 g
1 g
missing translation for 'unitSize'
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromopropane-1-D |
| CAS | 43217-00-1 |
| Molecular Formula | C3 D H6 Br |
| Quantity | 1 g |
| Synonym | Propane-1-d, 1-bromo- (6 CI, 9 CI), 1-Bromopropane-1-d1, 1-Bromopropane-1-d, 1-Bromopropane-d1, 1-Bromopropane-1-D |
| InChI Formula | InChI=1 S/C3H7Br/c1-2-3-4/h2-3H2,1H3/i3D |
| SMILES | [2 H]C(Br)CC |
| IUPAC Name | 1-bromo-1-deuteriopropane |
| Molecular Weight (g/mol) | 123.9978 |
| Formula Weight | 122.9794 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction