missing translation for 'onlineSavingsMsg'
Learn More

1-Bromotetradecane, 98%

Product Code. 10489680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10489680 500 g Glass bottle 500g
10377700 100 g Glass bottle 100g
13488418 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10489680 Supplier Thermo Scientific Acros Supplier No. 187385000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-71-0 | C14H29Br | 277.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-71-0
Molecular Formula C14H29Br
Molecular Weight (g/mol) 277.28
MDL Number MFCD00000228
InChI Key KOFZTCSTGIWCQG-UHFFFAOYSA-N
Synonym myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo
PubChem CID 8208
IUPAC Name 1-bromotetradecane
SMILES CCCCCCCCCCCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 5.0°C to 6.0°C
Color Yellow
Density 0.9300g/mL
Boiling Point 175.0°C to 178.0°C (20.0 mmHg)
Flash Point 149°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)13Br
Refractive Index 1.4590 to 1.4610
Quantity 500 g
Beilstein 01,II,136
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble
Formula Weight 277.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromotetradecane, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.