missing translation for 'onlineSavingsMsg'
Learn More

1-Bromotetradecane, 98%

Product Code. 11403097
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403097 2500 g 2500g
11483087 100 g 100g
11493087 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403097 Supplier Thermo Scientific Alfa Aesar Supplier No. A16082.0E
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-71-0 | C14H29Br | 277.29 g/mol

1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4?-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4′-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-71-0
Molecular Formula C14H29Br
Molecular Weight (g/mol) 277.29
MDL Number MFCD00000228
InChI Key KOFZTCSTGIWCQG-UHFFFAOYSA-N
Synonym myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo
PubChem CID 8208
IUPAC Name 1-bromotetradecane
SMILES CCCCCCCCCCCCCCBr

Specifications

Melting Point 4°C to 6°C
Density 1.018
Boiling Point 175°C to 178°C (20 mmHg)
Flash Point 158°C (316°F)
Refractive Index 1.46
Quantity 2500 g
Beilstein 1742640
Solubility Information Not miscible in water.
Formula Weight 277.3
Percent Purity 98%
Chemical Name or Material 1-Bromotetradecane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.