missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromoundecane-d23, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Bromoundecane-D23 |
| CAS | 349553-92-0 |
| Molecular Formula | C11 D23 Br |
| Quantity | 0.1 g |
| Synonym | Undecane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-d23, 11-bromo- (9 CI), 11-Bromoundecane-d23, 1-Bromoundecane-d23, 1-Bromoundecane-D23, 1-Undecyl-d23 bromide |
| InChI Formula | InChI=1 S/C11H23Br/c1-2-3-4-5-6-7-8-9-10-11-12/h2-11H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-tricosadeuterioundecane |
| Molecular Weight (g/mol) | 258.346 |
| Formula Weight | 257.2427 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction