missing translation for 'onlineSavingsMsg'
Learn More

1-Butanesulfonyl chloride, 98%

Product Code. 10299350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10299350 25 g Glass bottle 25g
15750869 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10299350 Supplier Thermo Scientific Acros Supplier No. 403320250
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2386-60-9 | C4H9ClO2S | 156.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-60-9
Molecular Formula C4H9ClO2S
Molecular Weight (g/mol) 156.63
MDL Number MFCD00007464
InChI Key WEDIIKBPDQQQJU-UHFFFAOYSA-N
Synonym 1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride
PubChem CID 75430
IUPAC Name butane-1-sulfonyl chloride
SMILES CCCCS(=O)(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow or Orange
Density 1.2000g/mL
Boiling Point 96.0°C (21.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3SO2Cl
Refractive Index 1.4540 to 1.4565
Quantity 25 g
Specific Gravity 1.2
Solubility Information Solubility in water: decomposes.
Formula Weight 156.63
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Butanesulfonyl chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Butanesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.