missing translation for 'onlineSavingsMsg'
Learn More

1-Butanethiol, 98%, AcroSeal™, Thermo Scientific™

Product Code. 10747518 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
Product Code. Quantity Packaging unitSize
10747518 800 mL AcroSeal™ Glass Bottle 800mL
10412098 100 mL AcroSeal™ Glass Bottle 100mL
2 options
This item is not returnable. View return policy

Product Code. 10747518

Brand: Thermo Scientific Acros 430878000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-79-5
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.18
MDL Number MFCD00004905
InChI Key WQAQPCDUOCURKW-UHFFFAOYSA-N
Synonym 1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
PubChem CID 8012
IUPAC Name butane-1-thiol
SMILES CCCCS

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -116.0°C
Color Colorless
Density 0.8400g/mL
Boiling Point 98.0°C
Flash Point 0.5°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3(CH2)3SH
Refractive Index 1.4420 to 1.4440
Quantity 800 mL
Merck Index 15,158
Specific Gravity 0.84
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 90.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Butanethiol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.