missing translation for 'onlineSavingsMsg'
Learn More

1-Butanethiol, 97%

Product Code. 10321274 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
5mL
This item is not returnable. View return policy

Product Code. 10321274

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-79-5 | C4H10S | 90.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-79-5
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.18
MDL Number MFCD00004905
InChI Key WQAQPCDUOCURKW-UHFFFAOYSA-N
Synonym 1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
PubChem CID 8012
IUPAC Name butane-1-thiol
SMILES CCCCS

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -116.0°C
Color Colorless
Density 0.8400g/mL
Boiling Point 98.0°C
Flash Point 0.5°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3(CH2)3SH
Refractive Index 1.4420 to 1.4440
Quantity 250 mL
Beilstein 01, 370
Fieser 02,53; 04,64
Merck Index 15, 1580
Specific Gravity 0.84
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 90.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1-Butanethiol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Butanethiol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.