missing translation for 'onlineSavingsMsg'
Learn More

1-Butanethiol, TRC

Product Code. 30344863
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30344863 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30344863

Brand: Toronto Research Chemicals TRCB5503055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Butanethiol
CAS 109-79-5
Molecular Formula C4 H10 S
Quantity 5 g
Synonym 1-Butyl mercaptan, 1-Mercaptobutane, Butanethiol, Butyl mercaptan, Butylthiol, Thiobutyl alcohol, n-Butane-1-thiol, n-Butanethiol, n-Butyl mercaptan, n-Butyl thioalcohol, n-Butylthiol
InChI Formula InChI=1S/C4H10S/c1-2-3-4-5/h5H,2-4H2,1H3
SMILES CCCCS
IUPAC Name butane-1-thiol
Molecular Weight (g/mol) 90.19
Formula Weight 90.0503
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.