missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, 99.9%, Solstice

Product Code. 15665800 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
20 L
4 x 2.5 L
6 x 1 L
Packaging:
Aluminum bottle
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
100mL
2.5 L
20 L
4 L
Pack of 4
Pack of 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15665800 6 x 1 L Glass Bottle Pack of 6
15608430 100 mL Aluminum bottle 4 L
15615800 100 mL Glass Bottle 100mL
15625800 1 L Stainless Steel Can 1 L
15635800 2.5 L Glass Bottle 2.5 L
15655800 4 x 2.5 L Glass Bottle Pack of 4
15645800 20 L Glass Bottle 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15665800 Supplier Fluka Supplier No. 5379936X1L
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

≥99.9%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point ≤-90°C
Color Colorless
Appearance Liquid
Density 0.81g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 11.2%
Ignition Point 649°C
Packaging Glass Bottle
Linear Formula CH3(CH2)3OH
UN Number UN1120
Beilstein 969148
Boiling Point 119°C
Molecular Weight (g/mol) 74.123
Formula Weight 74.12g/mol
Percent Purity 99%
Chemical Name or Material 1-Butanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.