missing translation for 'onlineSavingsMsg'
Learn More

1-Butoxy-4-eth-1-ynylbenzene, 97%, Thermo Scientific™

Product Code. 10572262
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10572262 1 g Amber Glass Bottle 1g
10282912 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10572262 Supplier Thermo Scientific Maybridge Supplier No. BTB09896DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 79887-15-3
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.24
MDL Number MFCD00173878
InChI Key JMLXBCSKFMMFGF-UHFFFAOYSA-N
Synonym 1-butoxy-4-eth-1-ynylbenzene, benzene,1-butoxy-4-ethynyl, 4-n-butoxyphenylacetylene, 4-butoxyphenylacetylene, 1-ethynyl-4-butoxybenzene, 1-butoxy-4-ethynyl-benzene
PubChem CID 2775120
IUPAC Name 1-butoxy-4-ethynylbenzene
SMILES CCCCOC1=CC=C(C=C1)C#C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3(CH2)3OC6H4Cidentical toCH
Quantity 1 g
Formula Weight 174.24
Percent Purity 97%
Chemical Name or Material 1-Butoxy-4-eth-1-ynylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.