missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium hexafluorophosphate, 98+%

Product Code. 10215821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10215821 25 g Glass bottle 25g
10612364 5 g Glass bottle 5g
11946261 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10215821 Supplier Thermo Scientific Acros Supplier No. 354170250
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 174501-64-5 | C8H15F6N2P | 284.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ionic liquid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174501-64-5
Molecular Formula C8H15F6N2P
Molecular Weight (g/mol) 284.19
MDL Number MFCD03093295
InChI Key IXQYBUDWDLYNMA-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium hexafluorophosphate, 1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, unii-zge3n4o8q9, 1-butyl-3-methylimidazoliumhexafluorophosphate, 1-n-butyl-3-methylimidazolium hexafluorophosphate, zge3n4o8q9, butylmethylimidazolium hexafluorophosphate, 1-methyl-3-butylimidazolium hexafluorophosphate, bmim pf6, c4mim hexafluorophosphate
PubChem CID 2734174
SMILES F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 6.5°C
Color Colorless to Yellow
Density 1.3727g/mL
Boiling Point >340.0°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 15,1584
Specific Gravity 1.3727
IUPAC Name 1-butyl-3-methylimidazol-3-ium;hexafluorophosphate
Viscosity 272.1 mPa.s (25°C)
Formula Weight 284.18
Percent Purity 98+%
Physical Form Viscous Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium hexafluorophosphate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.