missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-4-eth-1-ynylbenzene, 97%, Thermo Scientific™

Product Code. 10079191
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10079191 10 g 10g
10204644 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10079191 Supplier Thermo Scientific Maybridge Supplier No. BTB09904EA
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 79887-09-5
Molecular Formula C12H14
Molecular Weight (g/mol) 158.24
MDL Number MFCD00173885
InChI Key ZVWWYEHVIRMJIE-UHFFFAOYSA-N
Synonym 4-butylphenylacetylene, 1-butyl-4-eth-1-ynylbenzene, 1-butyl-4-eth1-ynylbenzene, 1-butyl-4-ethynyl-benzene, 4-n-butylphenylacetylene, p-butylphenylacetylene, benzene, 1-butyl-4-ethynyl, pubchem10978, acmc-209pic, p-butylphenyl acetylene
PubChem CID 2775132
IUPAC Name 1-butyl-4-ethynylbenzene
SMILES CCCCC1=CC=C(C=C1)C#C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Density 0.906g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula HC2CC6H4(CH2)3CH3
Quantity 10 g
Formula Weight 158.24
Percent Purity 97%
Chemical Name or Material 1-Butyl-4-eth-1-ynylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.