Learn More
Thermo Scientific™ Cartridges for Multidrop™ Pico Digital Dispensers
Product Code. 31722822 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
1 nL to 10 μL
11 pL to 10 μL
8 pL to 10 μL
Channels:
1
4
8
Model:
1-channel 200 μL Cartridges
1-channel 20 μL Cartridges
4-channel 200 μL Standard Cartridges
8-channel 20 μL Standard Cartridges
Surfactant Free Cartridges, 4-channel 200 μL
Surfactant Free Cartridges, 8-channel 20 μL
Unit Size:
200µL
20µL
Each
Pack of 20
Pack of 30
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Channels Model unitSize
31722822 8 pL to 10 μL 8 Surfactant Free Cartridges, 8-channel 20 μL 20µL
17961961 11 pL to 10 μL 1 1-channel 20 μL Cartridges Pack of 30
17971961 1 nL to 10 μL 1 1-channel 200 μL Cartridges Pack of 30
16643853 1 nL to 10 μL 4 4-channel 200 μL Standard Cartridges Each
17991961 1 nL to 10 μL 4 4-channel 200 μL Standard Cartridges Pack of 20
16653853 11 pL to 10 μL 8 8-channel 20 μL Standard Cartridges Each
17981961 11 pL to 10 μL 8 8-channel 20 μL Standard Cartridges Pack of 20
31722820 1 nL to 10 μL 4 Surfactant Free Cartridges, 4-channel 200 μL 200µL
31337545 1 nL to 10 μL 4 Surfactant Free Cartridges, 4-channel 200 μL -
31337544 8 pL to 10 μL 8 Surfactant Free Cartridges, 8-channel 20 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 31722822 Supplier Thermo Scientific™ Supplier No. LTR0006
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Cartridges for Multidrop™ Pico Digital Dispensers allow dispensing of one to eight fluids per cartridge, from 8 pL to 10 μL of DMSO, aqueous fluids, or master mixes, with or without small molecules or biomolecules.
The cartridges are designed for use with Multidrop™ Pico Digital Dispensers, dispensing volumes from 8 pL to 10 μL. Use the 1-channel cartridges with the Multidrop™ Pico 1, and the 4- or 8-channel standard or surfactant free cartridges with the Multidrop™ Pico 8 Surfactant Free (Pico 8 sf).
Features:
  • Single-Channel Cartridges: Use the 1-channel cartridges with the Pico 1 to dispense DMSO, aqueous fluids with added surfactant, or master mixes. Dispensing volume: 11 pL – 10 μL..
  • Standard Cartridges: Use the 4- or 8-channel standard cartridges with the Pico 8 sf to dispense DMSO, aqueous fluids with added surfactant, or master mixes. Dispensing volume: 11 pL – 10 μL.
  • Surfactant Free Cartridges: Use the 4- or 8-channel surfactant free cartridges with the Pico 8 sf to dispense only aqueous fluids without surfactant. Dispensing volume: 8 pL – 10 μL.
  • Contamination-Free Dispensing: Non-contact dispensing and single-use cartridges prevent carry-over and contamination.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Digital Dispenser Cartridge
Channels 8
Volume (Metric) 8 pL to 10 μL
For Use With (Equipment) Multidrop Pico 8 Surfactant Free Digital Dispenser
Model Surfactant Free Cartridges, 8-channel 20 μL
No. per Pack Case of 20