missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Chloro-1,1,2-trifluoro-2-iodoethane, 97%

Product Code. 11309365
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309365 5 g 5g
11319365 25 g 25g
11329365 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11309365 Supplier Alfa Aesar™ Supplier No. L16721.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 354-26-7
Molecular Formula C2HClF3I
Molecular Weight (g/mol) 244.38
MDL Number MFCD00042138
InChI Key XONGRFUEHPBOBB-UHFFFAOYSA-N
PubChem CID 78980
IUPAC Name 1-chloro-1,1,2-trifluoro-2-iodoethane
SMILES C(C(F)(F)Cl)(F)I

Specifications

Density 2.181
Boiling Point 77°C
Linear Formula ICHFCF2Cl
Refractive Index 1.432
Quantity 5 g
Beilstein 1737257
Sensitivity Light sensitive
Synonym 1-chloro-2-iodo-1,1,2-trifluoroethane, acmc-1cr56, 2-chloro-1,2,2-trifluoroethyl iodide, 1,2,2-trifluoro-2-chloro-1-iodoethane, ethane,1-chloro-1,1,2-trifluoro-2-iodo, ethane, 1-chloro-1,1,2-trifluoro-2-iodo
Formula Weight 244.38
Percent Purity 97%
Chemical Name or Material 1-Chloro-1,1,2-trifluoro-2-iodoethane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.