missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-fluoro-3-nitrobenzene, 97%

Product Code. 15418588
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418588 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15418588 Supplier Thermo Scientific Alfa Aesar Supplier No. H61989.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2106-49-2 | C6H3ClFNO2 | 175.54 g/mol

1-Chloro-2-fluoro-3-nitrobenzene is used as pharmaceutical intermediates, agrochemical intermediates, intermediates of medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-fluoro-3-nitrobenzene is used as pharmaceutical intermediates, agrochemical intermediates, intermediates of medicine.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing a
TRUSTED_SUSTAINABILITY

Specifications

CAS 2106-49-2
Color Yellow
Molecular Formula C6H3ClFNO2
MDL Number MFCD00069417
Quantity 25 g
UN Number UN2810
Synonym 3-chloro-2-fluoronitrobenzene, 2-fluoro-3-chloronitrobenzene, 1-chloro-2-fluoro-3-nitro-benzene, 2-fluoro-3-chloro-nitrobenzene, benzene, chlorofluoronitro, ccris 9050, 3-chloro-2-fluoro-1-nitrobenzene, pubchem2483, chlorofluoronitrobenzene, acmc-1bn8r
Solubility Information Soluble in water.
InChI Key RBAHXNSORRGCQA-UHFFFAOYSA-N
SMILES [O-][N+](=O)C1=CC=CC(Cl)=C1F
IUPAC Name 1-chloro-2-fluoro-3-nitrobenzene
Molecular Weight (g/mol) 175.54
PubChem CID 75016
Formula Weight 175.54
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-fluoro-3-nitrobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.