missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-methyl-2-propanol, 98%

Product Code. 11413218
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11413218

Brand: Thermo Scientific Alfa Aesar A18978.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 558-42-9 | C4H9ClO | 108.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 558-42-9
Molecular Formula C4H9ClO
Molecular Weight (g/mol) 108.57
MDL Number MFCD00021807
InChI Key JNOZGFXJZQXOSU-UHFFFAOYSA-N
Synonym 1-chloro-2-methyl-2-propanol, chloro-tert-butanol, 2-propanol, 1-chloro-2-methyl, 1-chloro-tert-butyl alcohol, 2-chloro-tert-butyl alcohol, 1-chloro-2-methyl-propan-2-ol, pubchem3675, 1-chloro-2-isobutanol, acmc-209lpr, chloro-tert-butyl alcohol
PubChem CID 68409
IUPAC Name 1-chloro-2-methylpropan-2-ol
SMILES CC(C)(O)CCl

Specifications

Density 1.058
Boiling Point 124°C to 128°C
Flash Point 38°C (100°F)
Refractive Index 1.44
Quantity 25 g
UN Number UN1987
Beilstein 1697054
Formula Weight 108.57
Percent Purity 98%
Chemical Name or Material 1-Chloro-2-methyl-2-propanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.