missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-methylpropane, 98%

Product Code. 11376826
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376826 50 g 50g
11386826 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376826 Supplier Thermo Scientific Alfa Aesar Supplier No. L04181.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 513-36-0 | C4H9Cl | 92.566 g/mol

1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

Solubility
Miscible with water, alcohol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-36-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000954
InChI Key QTBFPMKWQKYFLR-UHFFFAOYSA-N
PubChem CID 10554
IUPAC Name 1-chloro-2-methylpropane
SMILES CC(C)CCl

Specifications

Melting Point -131°C
Density 0.879
Boiling Point 68°C to 69°C
Flash Point −7°C (19°F)
Odor Sweet
Refractive Index 1.3975
Quantity 50 g
UN Number UN1127
Beilstein 635650
Merck Index 14,5138
Synonym isobutyl chloride, propane, 1-chloro-2-methyl, 1-chloro-2-methyl-propane, isobutylchlorid, iso-c4h9cl, sjphadikup@, acmc-1akdy, propane,1-chloro-2-methyl, ksc491k9j, 1-chloro-2-methylpropane
Solubility Information Miscible with water,alcohol and diethyl ether.
Formula Weight 92.57
Percent Purity 98%
Chemical Name or Material 1-Chloro-2-methylpropane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.