missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-propanol (70%, Technical Grade), TRC

Product Code. 30360943
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360943 2.5 g 2.5g
30357707 5 g -
30360736 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30360943 Supplier Toronto Research Chemicals Supplier No. TRCC5855802.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-00-4
Molecular Formula C3 H7 Cl O
Molecular Weight (g/mol) 94.54
Synonym 1-Chloro-2-propanol, (±)-1-Chloro-2-propanol, 1-Chloro-2-hydroxypropane, 1-Chloroisopropyl alcohol, NSC 77373, sec-Propylene chlorohydrin, α-Propylene chlorohydrin
IUPAC Name 1-chloropropan-2-ol
SMILES CC(O)CCl

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C3H7ClO/c1-3(5)2-4/h3,5H,2H2,1H3
Formula Weight 94.0185
Chemical Name or Material 1-Chloro-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.