missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-iodopropane, 98%, stabilized

Product Code. 10523961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10523961 100 g Glass bottle 100g
10114510 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10523961 Supplier Thermo Scientific Acros Supplier No. 215441000
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6940-76-7 | C3H6ClI | 204.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6940-76-7
Molecular Formula C3H6ClI
Molecular Weight (g/mol) 204.44
MDL Number MFCD00001096
InChI Key SFOYQZYQTQDRIY-UHFFFAOYSA-N
Synonym propane, 1-chloro-3-iodo, 3-chloropropyl iodide, trimethylene chloroiodide, 1-chloro-3-iodo-propane, 3-chloroiodopropane, 3-chloro-iodopropane, 1-chloro-3iodopropane, 1-choro-3-iodopropane, 3-chloro-l-iodopropane, 1-iodo-3-chloropropane
PubChem CID 81363
IUPAC Name 1-chloro-3-iodopropane
SMILES C(CCl)CI

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Orange to Pink or Yellow
Density 1.9000g/mL
Boiling Point 170.0°C to 172.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)3I
Refractive Index 1.5463 to 1.5483
Quantity 100 g
Beilstein 01, 114
Specific Gravity 1.9
Stabilizer copper
Formula Weight 204.44
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Chloro-3-iodopropane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Chloro-3-iodopropane, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.