missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-methylbutane, 98%

Product Code. 11397736
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397736 10 g 10g
11307746 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397736 Supplier Thermo Scientific Alfa Aesar Supplier No. L11371.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-84-6 | C5H11Cl | 106.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-84-6
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.593
MDL Number MFCD00013693
InChI Key CZHLPWNZCJEPJB-UHFFFAOYSA-N
Synonym isoamyl chloride, butane, 1-chloro-3-methyl, 3-methylbutyl chloride, isopentyl chloride, 4-chloro-2-methylbutane, 1-chloro-3,3-dimethylpropane, unii-87d3zl9f7a, isoamylchlorid, acmc-2098xt, 1-chloro-3-methyl-butane
PubChem CID 7893
IUPAC Name 1-chloro-3-methylbutane
SMILES CC(C)CCCl

Specifications

Melting Point -104°C
Boiling Point 99°C to 100°C
Flash Point 16°C (60°F)
Quantity 10 g
UN Number UN1107
Beilstein 1730988
Merck Index 14,5116
Formula Weight 106.6
Percent Purity 98%
Chemical Name or Material 1-Chloro-3-methylbutane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.