missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-nitrobenzene, 98%

Product Code. 11465397
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465397 250 g 250g
11455397 50 g 50g
11445397 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465397 Supplier Thermo Scientific Alfa Aesar Supplier No. B23423.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121-73-3 | C6H4ClNO2 | 157.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-73-3
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.55
MDL Number MFCD00007202
InChI Key KMAQZIILEGKYQZ-UHFFFAOYSA-N
Synonym m-chloronitrobenzene, 3-nitrochlorobenzene, 3-chloronitrobenzene, m-nitrochlorobenzene, benzene, 1-chloro-3-nitro, chloro-m-nitrobenzene, 3-chloro-1-nitrobenzene, metachloronitrobenzene, 1-chloronitrobenzene, 1-nitro-3-chlorobenzene
PubChem CID 8489
ChEBI CHEBI:82420
IUPAC Name 1-chloro-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(Cl)=C1

Specifications

Melting Point 43°C to 47°C
Density 1.34
Boiling Point 235°C to 236°C
Flash Point 124°C (255°F)
Quantity 250 g
UN Number UN1578
Beilstein 1907947
Merck Index 14,2151
Formula Weight 157.56
Percent Purity 98%
Chemical Name or Material 1-Chloro-3-nitrobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.