missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-ethylbenzene, 97%, Thermo Scientific Chemicals

Product Code. 11302107
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11302107 5 g 5g
11312107 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302107 Supplier Thermo Scientific Alfa Aesar Supplier No. L07931.06
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 622-98-0 | Molecular Formula: C8H9Cl | Molecular Weight: 140.61 g/mol

1-Chloro-4-ethylbenzene is used in paint & coating industry, rubber industry, water-treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-4-ethylbenzene is used in paint & coating industry, rubber industry, water-treatment.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-98-0
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00040855
InChI Key GPOFSFLJOIAMSA-UHFFFAOYSA-N
Synonym benzene, 1-chloro-4-ethyl, p-chloroethylbenzene, 4-chloroethylbenzene, 4-ethylchlorobenzene, acmc-20aovw, p-chlorophenyl ethane, 4-chloro ethylbenzene, 1-ethyl-4-chlorobenzene, 4-chloro-1-ethylbenzene, 1-chloro-4-ethyl benzene
PubChem CID 69330
IUPAC Name 1-chloro-4-ethylbenzene
SMILES CCC1=CC=C(C=C1)Cl

Specifications

Density 1.03
Quantity 5 g
Odor Aromatic
Refractive Index 1.5175
Beilstein 1099335
Boiling Point 184°C to 185°C
Solubility Information Difficult to mix.
Formula Weight 140.61
Percent Purity 97%
Chemical Name or Material 1-Chloro-4-ethylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.