missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-nitrobenzene, 98+%

Product Code. 11479150
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479150 250 g 250g
11459150 1000 g 1000g
11469150 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479150 Supplier Thermo Scientific Alfa Aesar Supplier No. A15396.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-00-5 | C6H4ClNO2 | 157.553 g/mol

1-Chloro-4-nitrobenzene is an important intermediate used in the preparation of 2,4-dinitrochlorobenzene and 3,4-dichloronitrobenzene. It is used as a raw material in the preparation of 4-nitrodiphenylamine, 4-chloroaninline and (4-[(4-aminobenzene)sulfonyl]aniline). It serves as a common antioxidant, which is found in rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-4-nitrobenzene is an important intermediate used in the preparation of 2,4-dinitrochlorobenzene and 3,4-dichloronitrobenzene. It is used as a raw material in the preparation of 4-nitrodiphenylamine, 4-chloroaninline and (4-[(4-aminobenzene)sulfonyl]aniline). It serves as a common antioxidant, which is found in rubber.

Solubility
Soluble in boiling hot ethanol, ether, toluene, acetone, organic solvents and carbon disulfide. Sparingly soluble in cold alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-00-5
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD00007285
InChI Key CZGCEKJOLUNIFY-UHFFFAOYSA-N
Synonym 4-chloronitrobenzene, p-chloronitrobenzene, p-nitrochlorobenzene, benzene, 1-chloro-4-nitro, 4-nitrochlorobenzene, pncb, 4-chloro-1-nitrobenzene, p-nitrophenyl chloride, p-nitrochlorobenzol, p-nitroclorobenzene
PubChem CID 7474
ChEBI CHEBI:34399
IUPAC Name 1-chloro-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])Cl

Specifications

Melting Point 82°C to 86°C
Density 1.515
Boiling Point 238°C to 242°C
Flash Point 127°C (260°F)
Quantity 250 g
UN Number UN1578
Beilstein 508691
Merck Index 14,2151
Solubility Information Soluble in boiling hot ethanol,ether,toluene,acetone,organic solvents and carbon disulfide. Sparingly soluble in cold alcohol. Insoluble in water.
Formula Weight 157.56
Percent Purity ≥98%
Chemical Name or Material 1-Chloro-4-nitrobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.