missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-nitrobenzene, 99%

Product Code. 10356360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
1kg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10356360 5 g Glass Bottle 5g
10366360 100 g Plastic bottle 100g
10646292 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10356360 Supplier Thermo Scientific Acros Supplier No. 109630050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 100-00-5 | C6H4ClNO2 | 157.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-00-5
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.56
MDL Number MFCD00007285
InChI Key CZGCEKJOLUNIFY-UHFFFAOYSA-N
Synonym 4-chloronitrobenzene, p-chloronitrobenzene, p-nitrochlorobenzene, benzene, 1-chloro-4-nitro, 4-nitrochlorobenzene, pncb, 4-chloro-1-nitrobenzene, p-nitrophenyl chloride, p-nitrochlorobenzol, p-nitroclorobenzene
PubChem CID 7474
ChEBI CHEBI:34399
IUPAC Name 1-chloro-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 81.0°C to 84.0°C
Color Yellow
Density 1.2980g/mL
Boiling Point 242.0°C
Flash Point 127°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula ClC6H4NO2
Quantity 5 g
Beilstein 05, 243
Merck Index 15, 2153
Specific Gravity 1.298
Solubility Information Solubility in water: insoluble.,soluble in organic solvents
Formula Weight 157.56
Percent Purity 99%
Physical Form Crystals or Flakes
Chemical Name or Material 1-Chloro-4-nitrobenzene, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Chloro-4-nitrobenzene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.