missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-propoxy-9H-thioxanthen-9-one, TRC

Product Code. 30012644
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012644 250 mg 250mg
30005565 500 mg 500mg
30010993 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012644 Supplier Toronto Research Chemicals Supplier No. TRCC424838250MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142770-42-1
Molecular Formula C16 H13 Cl O2 S
Molecular Weight (g/mol) 304.79
Synonym 1-Chloro-4-propoxy-9H-thioxanthen-9-one, 1-Chloro-4-propoxy-9H-thioxanthone, 1-Chloro-4-propoxythioxanthone, CPTX, Kayacure CPTX, Quantacure CPTX, Runtecure 1108, Speedcure CPTX
IUPAC Name 1-chloro-4-propoxythioxanthen-9-one
SMILES CCCOc1ccc(Cl)c2C(=O)c3ccccc3Sc12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C16H13ClO2S/c1-2-9-19-12-8-7-11(17)14-15(18)10-5-3-4-6-13(10)20-16(12)14/h3-8H,2,9H2,1H3
Formula Weight 304.0325
Chemical Name or Material 1-Chloro-4-propoxythioxanthone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.