missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-5-iodopentane, 97%, Thermo Scientific Chemicals

Product Code. 11311597
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311597 10 g 10g
11321597 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311597 Supplier Thermo Scientific Alfa Aesar Supplier No. L04469.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Chloro-5-iodopentane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-5-iodopentane is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60274-60-4
Molecular Formula C5H10ClI
Molecular Weight (g/mol) 232.489
MDL Number MFCD00039416
InChI Key GUUHKOCYWBEGGX-UHFFFAOYSA-N
PubChem CID 108914
IUPAC Name 1-chloro-5-iodopentane
SMILES C(CCCl)CCI

Specifications

Density 1.663
Boiling Point 75°C to 77°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5305
Quantity 10 g
Beilstein 1732342
Sensitivity Light sensitive
Synonym pentane, 1-chloro-5-iodo, pentamethylene chloroiodide, 5-chloropentyl iodide, 5-iodopentyl chloride, l-chloro-5-iodopentane, 1-iodo-5-chloropentane, 1-chloro-5-iodo-pentane, pentane,1-chloro-5-iodo, acmc-1b36d, 1-chloro-5-iodopentane
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 232.5
Percent Purity 97%
Chemical Name or Material 1-Chloro-5-iodopentane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.