missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-6-iodohexane, 97%

Product Code. 11391847
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11391847 50 g 50g
11381847 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391847 Supplier Thermo Scientific Alfa Aesar Supplier No. L05198.18
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34683-73-3 | C6H12ClI | 246.516 g/mol

1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store at -20°C. Light Sensitive. Store away from oxidizing agents. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34683-73-3
Molecular Formula C6H12ClI
Molecular Weight (g/mol) 246.516
MDL Number MFCD00039417
InChI Key QTJHNJCILMMRIQ-UHFFFAOYSA-N
Synonym hexane, 1-chloro-6-iodo, hexamethylene chloroiodide, 6-chlorohexyl iodide, acmc-20aovr, 6-chloro-1-iodohexane, 1-iodo-6-chloro hexane, 1-chloro-6-iodohexane, 1-chloro-6-iodohexane, stab. with copper chip, stabilized with copper chip
PubChem CID 118713
IUPAC Name 1-chloro-6-iodohexane
SMILES C(CCCI)CCCl

Specifications

Refractive Index 1.525
Quantity 50 g
Beilstein 1732408
Sensitivity Light sensitive
Solubility Information Not miscible in water.
Formula Weight 246.52
Percent Purity 97%
Chemical Name or Material 1-Chloro-6-iodohexane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.