missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroacetyl-2-(trifluoroacetyl)hydrazine, 97%

Product Code. 15451436
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15451436 250 mg 250mg
15490878 1 g 1g
15400888 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451436 Supplier Thermo Scientific Alfa Aesar Supplier No. H54647.MD
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 762240-99-3 | C4H4ClF3N2O2 | 204.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762240-99-3
Molecular Formula C4H4ClF3N2O2
Molecular Weight (g/mol) 204.53
MDL Number MFCD07779449
InChI Key DYKIVKLXFDNBMY-UHFFFAOYSA-N
Synonym 1-chloroacetyl-2-trifluoroacetyl hydrazine, n'-2-chloroacetyl-2,2,2-trifluoroacetohydrazide, n'-chloroacetyl-2,2,2-trifluoroacetohydrazide, acetic acid, 2,2,2-trifluoro-, 2-2-chloroacetyl hydrazide, 2-chloro-n'-2,2,2-trifluoroacetyl acetohydrazide, n'-2-chloroacetyl-trifluoroacetohydrazide, 1-trifluoroacetyl-2-chloroacetyl hydrazine, 2-chloro-n'-trifluoroacetyl acetohydrazide, acetic acid, trifluoro-, 2-chloroacetyl hydrazide
PubChem CID 11961370
IUPAC Name N'-(2-chloroacetyl)-2,2,2-trifluoroacetohydrazide
SMILES FC(F)(F)C(=O)NNC(=O)CCl

Specifications

Quantity 250 mg
Formula Weight 204.53
Percent Purity 97%
Chemical Name or Material 1-Chloroacetyl-2-(trifluoroacetyl)hydrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.