missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroadamantane, 98%

Product Code. 15431077
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431077 100 g 100g
11307108 5 g 5g
11397098 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15431077 Supplier Thermo Scientific Alfa Aesar Supplier No. 015169.22
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 935-56-8 | C10H15Cl | 170.68 g/mol

It is used in some dry etching masks and polymer formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 935-56-8
Molecular Formula C10H15Cl
Molecular Weight (g/mol) 170.68
MDL Number MFCD00075627
InChI Key OZNXTQSXSHODFR-UHFFFAOYSA-N
Synonym 1-adamantyl chloride, adamantyl chloride, 3-chloroadamantane, adamantane, 1-chloro, tricyclo 3.3.1.13,7 decane, 1-chloro, 1-chlorotricyclo 3.3.1.13,7 decane, 3r,5s,7s-1-chloroadamantane, 1-chlorotricyclo 3.3.1.1∼3,7∼ decane
PubChem CID 64154
IUPAC Name 1-chloroadamantane
SMILES ClC12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 165°C
Quantity 100 g
Solubility Information Poorly soluble in water. Soluble in hydrocarbons. But readily soluble in nonpolar organic solvents.
Formula Weight 170.68
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-Chloroadamantane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.