missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroanthraquinone, 98%, Thermo Scientific™

Product Code. 10122470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10122470 5 g 5g
10449400 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10122470 Supplier Thermo Scientific Acros Supplier No. 108600050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 82-44-0
MDL Number MFCD00001189
InChI Key BOCJQSFSGAZAPQ-UHFFFAOYSA-N
Synonym 1-chloroanthraquinone, 1-chloro anthraquinone, anthraquinone, 1-chloro, 1-chloro-9,10-anthraquinone, 9,10-anthracenedione, 1-chloro, 1-chloranthrachinon, 1-chloranthrachinon czech, alpha-chloroanthraquinone, alpha-monochloroanthraquinone, unii-96s5sqv15v
PubChem CID 6709
IUPAC Name 1-chloroanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 157°C to 162°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C14H7ClO2
Quantity 5 g
Beilstein 07,787
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 242.66
Formula Weight 242.66
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 1-Chloroanthraquinone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.