missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorobutane, 99+%, pure

Product Code. 10478310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10478310 1 L Glass bottle 1 L
10193390 2.5 L Glass bottle 2.5 L
10627934 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10478310 Supplier Thermo Scientific Acros Supplier No. 154630010
  • ONLINE EXCLUSIVE PRICE
    53.40 €
    51.80 € / 1 L
    Save 1.60 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 109-69-3 | C4H9Cl | 92.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -123.0°C
Color Colorless
Density 0.8800g/mL
Boiling Point 77.0°C to 78.0°C
Flash Point −12°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3Cl
Refractive Index 1.4014 to 1.4034
Quantity 1 L
Beilstein 01, 118
Merck Index 15, 1561
Specific Gravity 0.88
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: miscible with alcohol and ether
Viscosity 0.45 mPa.s (20°C)
Formula Weight 92.57
Percent Purity 99+%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1-Chlorobutane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chlorobutane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H412-Harmful to aquatic life with long lasting effects.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.