missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorobutane CHROMASOLV™, for HPLC, ≥99.8%, Solstice

Product Code. 15614730
Change view
Click to view available options
Quantity:
1 L
2 L
2.5 L
Unit Size:
1 L
2 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15614730 2 L 2 L
15694720 1 L 1 L
15604730 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15614730 Supplier Honeywell-Chromasolv Supplier No. 349582L
  • ONLINE EXCLUSIVE PRICE
    259.00 €
    242.24 € / 2 L
    Save 16.76 €
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™, for HPLC,≥99.8%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

Melting Point -123°C
Density 0.886g/mL at 25°C (lit.)
Boiling Point 77°C to 78°C
Linear Formula CH3(CH2)3Cl
Refractive Index n20/D 1.402
Quantity 2 L
Beilstein 1730909
Formula Weight 92.57g/mol
Vapor Pressure 80.1mmHg (78.4°C)
Percent Purity ≥99.8%
Grade CHROMASOLV
Physical Form Liquid
Chemical Name or Material 1-Chlorobutane, for HPLC
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chlorobutane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.