missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorobutane, for Protein Sequencing, Fisher Chemical

Product Code. 10080050 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10080050 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10080050 Supplier Fisher Chemical Supplier No. C/4758/17
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 109-69-3
Melting Point -123°C
Color Colorless
Boiling Point 77°C
Packaging Amber glass bottle
Molecular Formula C4H9Cl
MDL Number 1009
Quantity 2.5 L
Formula Weight 92.57g/mol
Vapor Pressure 108mbar at 20°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chlorobutane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H412-Harmful to aquatic life with long lasting effects.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.