missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorobutane, ReagentPlus™, 99%, Honeywell™

Product Code. 15631150 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15631150 2.5 L Glass Bottle 2.5 L
15621150 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15631150 Proveedor Honeywell Research Chemicals N.º de proveedor 1250082.5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Especificaciones

Melting Point -123°C
Color Colorless
Density 0.886g/mL at 25°C
Boiling Point 78.8°C
Explosion Limit 0.101
Flash Point −12°C (10.4°F)
Ignition Point 860°F
Packaging Glass Bottle
Linear Formula CH3(CH2)3Cl
Refractive Index n20/D 1.402(lit.)
Quantity 2.5 L
UN Number UN1127
Beilstein 1730909
Formula Weight 92.57g/mol
Vapor Pressure 80.1mmHg (78.4°C)
Percent Purity 99%
Grade ReagentPlus
Chemical Name or Material 1-Chlorobutane
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.