missing translation for 'onlineSavingsMsg'
Learn More

1-chlorobutane, Sequenation Grade

Product Code. 10015770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10015770 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10015770 Supplier Thermo Scientific Acros Supplier No. 404550010
  • ONLINE EXCLUSIVE PRICE
    90.65 €
    88.00 € / 1 L
    Save 2.65 €
    2% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 109-69-3 | C4H9Cl | 92.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

Acidity 0.0003 meq/g max.
CAS 109-69-3
Absorbance (1 cm cell vs HPLC-grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 270nm, 0.005 max. at 300nm, 0.01 max. at 250nm, 1 max. at 220nm, 1 max. at 230nm
Melting Point -123.0°C
Color Brown to White
Density 0.8800g/mL
Boiling Point 77.0°C to 78.0°C
Flash Point −12°C
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C4H9Cl
Linear Formula CH3(CH2)3Cl
Refractive Index 1.4014 to 1.4034
MDL Number MFCD00001009
Quantity 1 L
Beilstein 01, 118
Residue after Evaporation 2ppm max.
Merck Index 15, 1561
Specific Gravity 0.88
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: miscible with alcohol and ether
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
SMILES CCCCCl
IUPAC Name 1-chlorobutane
Molecular Weight (g/mol) 92.57
PubChem CID 8005
Viscosity 0.45 mPa.s (20°C)
Formula Weight 92.57
Percent Purity ≥99.5% (GC)
Grade Sequenation
Physical Form Powder
Chemical Name or Material 1-Chlorobutane
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • 1-Chlorobutane
Palabra de advertencia
  • Danger
Clasificación
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Indicaciones de peligro
  • H304-May be fatal if swallowed and enters airways.
  • H412-Harmful to aquatic life with long lasting effects.
  • H225-Highly flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.