missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorohexane, 95%

Product Code. 10186793 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10186793

Brand: Thermo Scientific Acros 404645000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 544-10-5 | C6H13Cl | 120.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-10-5
Molecular Formula C6H13Cl
Molecular Weight (g/mol) 120.62
MDL Number MFCD00001018
InChI Key MLRVZFYXUZQSRU-UHFFFAOYSA-N
Synonym chlorohexane, hexyl chloride, hexane, 1-chloro, n-hexyl chloride, 1-chloro-hexane, hexane, chloro, unii-r5l7i6o9nw, r5l7i6o9nw, triisobutenyl chloride, hexylchloride
PubChem CID 10992
IUPAC Name 1-chlorohexane
SMILES CCCCCCCl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 0.8700g/mL
Boiling Point 135.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5Cl
Refractive Index 1.4189 to 1.4209
Quantity 500 g
Beilstein 01, 143
Merck Index 15, 2145
Specific Gravity 0.87
Formula Weight 120.62
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-chlorohexane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chlorohexane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.