missing translation for 'onlineSavingsMsg'
Learn More

1-Chloromethyl-1H-1,2,4-triazole, hydrochloride, 98%, Thermo Scientific™

Product Code. 15395048 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15395048 5 g 5g
15385048 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15395048 Supplier Thermo Scientific Acros Supplier No. 456090050
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 84387-61-1
Molecular Formula C3H5Cl2N3
Molecular Weight (g/mol) 153.99
MDL Number MFCD00964947
InChI Key GXVWDLWTFIDZLR-UHFFFAOYSA-N
Synonym 1-chloromethyl-1h-1,2,4-triazole hydrochloride, 1-chloromethyl-1,2,4-triazole hydrochloride, 1h-1,2,4-triazole,1-chloromethyl-,monohydrochloride, 1h-1,2,4-triazole, 1-chloromethyl-, monohydrochloride
PubChem CID 13268168
IUPAC Name 1-(chloromethyl)-1,2,4-triazole;hydrochloride
SMILES Cl.ClCN1C=NC=N1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 154
Percent Purity 98%
Chemical Name or Material 1-Chloromethyl-1H-1,2,4-triazole, hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.