missing translation for 'onlineSavingsMsg'
Learn More

1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate), 96%

Product Code. 10561881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10561881 5 g Glass bottle 5g
10202322 25 g Glass bottle 25g
11926531 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10561881 Supplier Thermo Scientific Acros Supplier No. 291490050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 140681-55-6 | C7H14B2ClF9N2 | 354.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140681-55-6
Molecular Formula C7H14B2ClF9N2
Molecular Weight (g/mol) 354.26
MDL Number MFCD00142607
InChI Key TXRPHPUGYLSHCX-UHFFFAOYSA-N
Synonym selectfluor, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane bis tetrafluoroborate, f-teda-bf4, 1-chloromethyl-4-fluoro-1,4-diazabicyclo 2.2.2 octane-1,4-diium tetrafluoroborate, unii-4p1za6r76d, selectfluor fluorinating reagent, f-teda, selectfluor r fluorinating reagent, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane ditetrafluoroborate
PubChem CID 2724933
IUPAC Name 1-(chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane;ditetrafluoroborate
SMILES F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2

Specifications

Melting Point 260.0°C
Color White
Flash Point >54°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Iodometry)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,8562
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hcl,moderately soluble in ch3cn,dmf,slightly soluble in (ch3)2co
Formula Weight 354.26
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.